Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_013796.3 | NP_038824.2 | 10 | 1402 | |
GenBank mRNA | AK032158.1 | - | 10 | 1403 | |
AK087468.1 | - | 10 | 1405 | ||
AK138035.1 | - | 10 | 1395 | ||
AK162633.1 | - | 10 | 1385 | ||
AK168830.1 | - | 9 | 1487 | ||
AK170219.1 | - | 10 | 1384 | ||
BC019640.1 | - | 9 | 1464 | ||
BC039790.1 | - | 10 | 1394 | 4951805 |