Gene Transcripts

Gene Symbol
Hspb7
Entrez Gene ID
29818
Gene Name
heat shock protein family, member 7 (cardiovascular)
Gene Aliases
27kDa, Hsp25-2, cvHsp
Chromosome Location
Chr. 4: 141420779 - 141425310 on Build GRCm38
UniGene ID
Mm.46181
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_013868.4 NP_038896.2 3 1801
GenBank mRNA AK030025.1 - 3 1802
AK086011.1 - 3 878
BC089584.1 - 3 876 6755510

View Full Product Details