Gene Transcripts

Gene Symbol
Snrpa
Entrez Gene ID
53607
Gene Name
small nuclear ribonucleoprotein polypeptide A
Gene Aliases
C430021M15Rik, Rnu1a-1, Rnu1a1, U1-A, U1A
Chromosome Location
Chr. 7: 27187006 - 27196271 on Build GRCm38
UniGene ID
Mm.386890
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001046637.1 NP_001040102.1 3 546
NM_001285825.1 NP_001272754.1 4 620
NM_015782.3 NP_056597.3 4 623
GenBank mRNA AK013886.1 - 4 597
AK088550.1 - 4 481
AK206903.1 - 2 81
BC003229.1 - 4 523 3483391
BC094006.1 - 4 483 4983854
BC096648.1 - 4 477 1397390
CX565356.1 - 3 516
DV650448.1 - 2 239
L15447.1 - 4 419

View Full Product Details