Gene Transcripts
- Gene Symbol
- SLC22A7
- Entrez Gene ID
- 10864
- Gene Name
- solute carrier family 22 member 7
- Gene Aliases
- NLT, OAT2
- Chromosome Location
- Chr. 6: 43295694 - 43305538 on Build GRCh38
- UniGene ID
- Hs.485438
- Species
- Homo sapiens
- Gene Symbol
- CRIP3
- Entrez Gene ID
- 401262
- Gene Name
- cysteine rich protein 3
- Gene Aliases
- TLP, TLP-A, bA480N24.2
- Chromosome Location
- Chr. 6: 43305473 - 43310705 on Build GRCh38
- UniGene ID
- Hs.653165
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AI042209.1 |
- |
1 |
41 |
|
View Full Product Details