Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001163704.1 | NP_001157176.1 | 6 | 1321 | |
NM_001163705.1 | NP_001157177.1 | 6 | 1105 | ||
NM_001163706.1 | NP_001157178.1 | 6 | 1097 | ||
NM_001163707.1 | NP_001157179.1 | 6 | 1132 | ||
NM_015797.4 | NP_056612.1 | 6 | 1847 | ||
GenBank mRNA | AF176526.1 | - | 6 | 1220 | |
AK002840.1 | - | 6 | 1057 | ||
AK144715.1 | - | 6 | 1261 | ||
AK150612.1 | - | 6 | 1289 | ||
AK152888.1 | - | 6 | 1171 | ||
AK165867.1 | - | 6 | 1832 | ||
AK170743.1 | - | 6 | 1098 | ||
AK171483.1 | - | 6 | 1098 | ||
AK179920.1 | - | 1 | 193 | ||
AK199261.1 | - | 1 | 193 | ||
BC017512.1 | - | 6 | 1117 | 4485218 |