Gene Transcripts

Gene Symbol
Uevld
Entrez Gene ID
54122
Gene Name
UEV and lactate/malate dehyrogenase domains
Gene Aliases
8430408E05Rik, Attp
Chromosome Location
Chr. 7: 46923216 - 46958530 on Build GRCm38
UniGene ID
Mm.1999
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001040695.1 NP_001035785.1 3 261
GenBank mRNA AK086750.1 - 1 996
AK154536.1 - 3 212
AK155685.1 - 3 208
BC054796.1 - 3 267 6843964
CB524764.1 - 3 267
CN538811.1 - 2 131
U97571.1 - 3 215

View Full Product Details