Gene Transcripts

Gene Symbol
Rragc
Entrez Gene ID
54170
Gene Name
Ras-related GTP binding C
Gene Aliases
AU041672, Gtr2, RAGC, TIB929, YGR163W
Chromosome Location
Chr. 4: 123917433 - 123936997 on Build GRCm38
UniGene ID
Mm.220922
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017475.2 NP_059503.2 6, 7 1154
GenBank mRNA AB017616.1 - 6, 7 1077
AK150792.1 - 6, 7 1109
AK159355.1 - 6, 7 1087
AK159434.1 - 6, 7 1109
AK166658.1 - 6, 7 1100
BC005417.1 - 6, 7 1102 3495022
BC037732.1 - 3, 4 451
BC071245.1 - 6, 7 1502

View Full Product Details