Gene Transcripts

Gene Symbol
Eif3d
Entrez Gene ID
55944
Gene Name
eukaryotic translation initiation factor 3, subunit D
Gene Aliases
66/67kDa, AA407891, Eif3s7, eIF3p66
Chromosome Location
Chr. 15: 77958998 - 77971088 on Build GRCm38
UniGene ID
Mm.3955
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018749.2 NP_061219.2 2 229
GenBank mRNA AB012580.1 - 2 173
AK045228.1 - 2 171
AK049767.1 - 2 168
AK145689.1 - 2 184
AK158344.1 - 2 203
AK168256.1 - 2 219
BC018598.1 - 2 167
BC089020.1 - 2 163 30551991

View Full Product Details