Gene Transcripts

Gene Symbol
Pcnx
Entrez Gene ID
54604
Gene Name
pecanex homolog (Drosophila)
Gene Aliases
2900024E21Rik, 3526401J03Rik, AF096286, AI327143, AI413187, Pcnxl1
Chromosome Location
Chr. 12: 81860028 - 82000924 on Build GRCm38
UniGene ID
Mm.86584
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018814.3 NP_061284.2 36 7784
GenBank mRNA AF096286.1 - 30 5614
AK014387.1 - 10 2472
AK129221.1 - 31 5664
BC066849.1 - 31 5447
BC098370.1 - 31 5374

View Full Product Details