Gene Transcripts

Gene Symbol
Fnbp4
Entrez Gene ID
55935
Gene Name
formin binding protein 4
Gene Aliases
FBP30, Fnbp30, mKIAA1014
Chromosome Location
Chr. 2: 90745370 - 90781021 on Build GRCm38
UniGene ID
Mm.489638
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018828.2 NP_061298.1 11, 12 2077
GenBank mRNA AK084018.1 - 11, 12 1869
U40750.2 - 11, 12 2077

View Full Product Details