Gene Transcripts

Gene Symbol
Egfl6
Entrez Gene ID
54156
Gene Name
EGF-like-domain, multiple 6
Gene Aliases
Maeg
Chromosome Location
Chr. X: 166523007 - 166585716 on Build GRCm38
UniGene ID
Mm.37707
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019397.3 NP_062270.1 9, 10 1368
GenBank mRNA AJ245672.1 - 9, 10 1370
AK078534.1 - 9, 10 1346
BC117702.2 - 9, 10 1354 40062530

View Full Product Details