Gene Transcripts

Gene Symbol
Irgm2
Entrez Gene ID
54396
Gene Name
immunity-related GTPase family M member 2
Gene Aliases
AI481100, GTPI, Ifggd2, Iigp2
Chromosome Location
Chr. 11: 58214977 - 58222783 on Build GRCm38
UniGene ID
Mm.458517
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019440.3 NP_062313.3 2 2674
GenBank mRNA AJ007972.1 - 1 2475
AK142291.1 - 2 2676
AK165347.1 - 2 2612

View Full Product Details