Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_019553.2 | NP_062426.2 | 2 | 663 | |
GenBank mRNA | AF159131.3 | - | 2 | 624 | |
AK010281.1 | - | 2 | 543 | ||
AK145578.1 | - | 2 | 647 | ||
AK160095.1 | - | 2 | 637 | ||
BC030895.1 | - | 1 | 95 | ||
BC043655.1 | - | 2 | 635 | 5363948 | |
BC059237.1 | - | 2 | 623 | 6412782 | |
BC060220.1 | - | 2 | 615 |