Gene Transcripts

Gene Symbol
Letm1
Entrez Gene ID
56384
Gene Name
leucine zipper-EF-hand containing transmembrane protein 1
Gene Aliases
Chromosome Location
Chr. 5: 33739675 - 33782704 on Build GRCm38
UniGene ID
Mm.260538
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019694.1 NP_062668.1 9 1525
GenBank mRNA AB041582.1 - 9 1538
AF061026.1 - 9 1525
AK133849.1 - 9 1545
AK161638.1 - 3 1247
AK218029.1 - 4 367
BC023862.1 - 9 1522
BC046326.1 - 9 1532
BC055865.1 - 9 1543
BC061115.1 - 9 1538 6533034
BC061690.1 - 9 1525

View Full Product Details