Gene Transcripts

Gene Symbol
Nupr1
Entrez Gene ID
56312
Gene Name
nuclear protein transcription regulator 1
Gene Aliases
2310032H04Rik, Com1, p8
Chromosome Location
Chr. 7: 126623246 - 126625470 on Build GRCm38
UniGene ID
Mm.18742
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019738.1 NP_062712.1 3 606
GenBank mRNA AF131196.1 - 3 606
AK009572.1 - 3 627
BC002109.1 - 3 612 3487434

View Full Product Details