Gene Transcripts

Gene Symbol
Akap8
Entrez Gene ID
56399
Gene Name
A kinase (PRKA) anchor protein 8
Gene Aliases
1200016A02Rik, AA673585, AKAP-8, AKAP95, AU015639
Chromosome Location
Chr. 17: 32303672 - 32321327 on Build GRCm38
UniGene ID
Mm.328945
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK077864.1 - 1 1381
AK089092.1 - 14 3404
BC042602.1 - 14 4274
BC060170.1 - 14 4271
BC060209.1 - 13 3887
BC078448.1 - 13 3777

View Full Product Details