Gene Transcripts
- Gene Symbol
- Snd1
- Entrez Gene ID
- 56463
- Gene Name
- staphylococcal nuclease and tudor domain containing 1
- Gene Aliases
- AL033314, Tudor-SN
- Chromosome Location
- Chr. 6: 28480315 - 28935162 on Build GRCm38
- UniGene ID
- Mm.439987
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_019776.2 |
NP_062750.2
|
13, 14 |
1681 |
|
GenBank mRNA |
AB021491.1 |
- |
13, 14 |
1638 |
|
AK034914.1 |
- |
12, 13 |
1550 |
|
AK088028.1 |
- |
13, 14 |
1640 |
|
AK133837.1 |
- |
13, 14 |
1687 |
|
AK159837.1 |
- |
13, 14 |
1646 |
|
AK161591.1 |
- |
13, 14 |
1635 |
|
AK162432.1 |
- |
13, 14 |
1622 |
|
AK167576.1 |
- |
13, 14 |
1671 |
|
BC007126.1 |
- |
13, 14 |
1628 |
3592548 |
View Full Product Details