Gene Transcripts

Gene Symbol
Aatf
Entrez Gene ID
56321
Gene Name
apoptosis antagonizing transcription factor
Gene Aliases
4933415H02Rik, 5830465M17Rik, Che-1, Trb
Chromosome Location
Chr. 11: 84422856 - 84513570 on Build GRCm38
UniGene ID
Mm.257482
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019816.1 NP_062790.1 4 704
GenBank mRNA AF222801.1 - 4 704
AK077789.1 - 4 728
AK141334.1 - 4 702
BC025080.1 - 4 691 2648117

View Full Product Details