Gene Transcripts

Gene Symbol
CENPI
Entrez Gene ID
2491
Gene Name
centromere protein I
Gene Aliases
CENP-I, FSHPRH1, LRPR1
Chromosome Location
Chr. X: 101098159 - 101181847 on Build GRCh38
UniGene ID
Hs.348920
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318521.1 NP_001305450.1 3, 4 410
NM_001318523.1 NP_001305452.1 1, 2 247
NM_006733.3 NP_006724.2 2, 3 504
XM_005262111.2 XP_005262168.1 3, 4 374
XM_011530894.2 XP_011529196.1 3, 4 403
XM_011530895.2 XP_011529197.1 4, 5 690
XM_011530897.2 XP_011529199.1 4, 5 565
XM_011530899.2 XP_011529201.1 2, 3 371
XM_017029383.1 XP_016884872.1 3, 4 403
GenBank mRNA AK302986.1 - 3, 4 403
BC012462.2 - 1, 2 248 4537558
DA736853.1 - 3, 4 410
X97249.1 - 2, 3 503

View Full Product Details