Gene Transcripts

Gene Symbol
Mllt11
Entrez Gene ID
56772
Gene Name
myeloid/lymphoid or mixed-lineage leukemia (trithorax homolog, Drosophila); translocated to, 11
Gene Aliases
AI839562, Af1q, Zfp692
Chromosome Location
Chr. 3: 95218544 - 95230459 on Build GRCm38
UniGene ID
Mm.331208
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019914.4 NP_064298.1 2 1364
GenBank mRNA AB000733.1 - 2 938
AK046005.1 - 2 868
AK152708.1 - 2 1364
AK158495.1 - 2 990
BC022963.1 - 2 805 5365628

View Full Product Details