Gene Transcripts

Gene Symbol
Chst5
Entrez Gene ID
56773
Gene Name
carbohydrate (N-acetylglucosamine 6-O) sulfotransferase 5
Gene Aliases
AI173964, GST-4, Gn6st-3, I-GlcNAc-6-ST
Chromosome Location
Chr. 8: 111889135 - 111910199 on Build GRCm38
UniGene ID
Mm.432506
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019950.2 NP_064334.1 2 588
GenBank mRNA AF176840.1 - 2 611

View Full Product Details