Gene Transcripts

Gene Symbol
Pik3cg
Entrez Gene ID
30955
Gene Name
phosphoinositide-3-kinase, catalytic, gamma polypeptide
Gene Aliases
5830428L06Rik, PI3Kgamma, p110gamma
Chromosome Location
Chr. 12: 32173129 - 32208677 on Build GRCm38
UniGene ID
Mm.101369
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001146200.1 NP_001139672.1 4 2389
NM_001146201.1 NP_001139673.1 4 2445
NM_020272.2 NP_064668.2 4 2566
GenBank mRNA AF208345.1 - 3 2245
AJ249280.1 - 4 2566
AK040543.1 - 4 2351
AK132553.1 - 4 2391
AK137470.1 - 4 2298
AK137620.1 - 4 2447
AK143238.1 - 1 70
BC051246.1 - 4 2306 6514979

View Full Product Details