Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199020.1 | NP_001185949.1 | 9 | 1417 | |
NM_021421.4 | NP_067396.3 | 8 | 1552 | ||
GenBank mRNA | AB041602.1 | - | 6 | 1085 | |
AK077039.1 | - | 8 | 1499 | ||
AK088834.1 | - | 7 | 1185 | ||
AK135709.1 | - | 9 | 1414 | ||
AK158731.1 | - | 8 | 1568 | ||
AK166619.1 | - | 9 | 1416 | ||
AK169653.1 | - | 7 | 1136 | ||
AK199712.1 | - | 2 | 63 | ||
AK214324.1 | - | 2 | 63 | ||
AK216183.1 | - | 2 | 63 | ||
BC024907.1 | - | 8 | 1512 | 2812124 |