Gene Transcripts

Gene Symbol
Rsrp1
Entrez Gene ID
27981
Gene Name
arginine/serine rich protein 1
Gene Aliases
2700043I21Rik, D4Ucla2, D4Wsu53e
Chromosome Location
Chr. 4: 134923625 - 134927370 on Build GRCm38
UniGene ID
Mm.331964
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AB041555.1 - 5 1047
AK003799.1 - 3 1618
AK028145.1 - 3 1620
AK029529.1 - 3 1616
AK160993.1 - 3 1618
AK169598.1 - 5 1071
AK169760.1 - 3 1618
BC043057.1 - 3 1602 6419597
BC046231.1 - 3 1519 6813388
BC054728.1 - 3 1590 6419723
BC056986.1 - 3 1551 6816649

View Full Product Details