Gene Transcripts

Gene Symbol
Nelfe
Entrez Gene ID
27632
Gene Name
negative elongation factor complex member E, Rdbp
Gene Aliases
D17H6S45, NELF-E, RD, Rdbp
Chromosome Location
Chr. 17: 34850391 - 34856372 on Build GRCm38
UniGene ID
Mm.279907
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001045863.1 NP_001039328.1 11 1267
NM_001045864.1 NP_001039329.1 11 1353
NM_138580.2 NP_613046.2 11 1141
GenBank mRNA AK011663.1 - 11 1126
AK012389.1 - 11 1158
AK131947.1 - 11 1125
AK147011.1 - 11 1249
BC034867.1 - 11 1093 1495547

View Full Product Details