Gene Transcripts

Gene Symbol
DBF4
Entrez Gene ID
10926
Gene Name
DBF4 zinc finger
Gene Aliases
ASK, CHIF, DBF4A, ZDBF1
Chromosome Location
Chr. 7: 87876229 - 87909541 on Build GRCh38
UniGene ID
Hs.485380
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318060.1 NP_001304989.1 10 1434
NM_001318061.1 NP_001304990.1 11 1449
NM_001318062.1 NP_001304991.1 11 1480
NM_006716.3 NP_006707.1 11 1541
GenBank mRNA AB028069.1 - 11 1554
AB028070.1 - 11 1493
AF160249.1 - 11 1253
AF160876.1 - 11 1236
AK292569.1 - 11 1283
AK302010.1 - 10 1063
AK303175.1 - 11 1145
BC036045.1 - 11 1347 5273059
BC047693.1 - 11 1222 5753567

View Full Product Details