Gene Transcripts

Gene Symbol
Traf6
Entrez Gene ID
22034
Gene Name
TNF receptor-associated factor 6
Gene Aliases
2310003F17Rik, AI851288, C630032O20Rik
Chromosome Location
Chr. 2: 101678420 - 101701668 on Build GRCm38
UniGene ID
Mm.292729
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AK041172.1 - 2 420
AK155434.1 - 2 390
BC060705.1 - 3 611 6409657
D84655.1 - 3 464

View Full Product Details