Gene Transcripts

Gene Symbol
Ttk
Entrez Gene ID
22137
Gene Name
Ttk protein kinase
Gene Aliases
AL022661, Esk1, Mps1, PYT, esk
Chromosome Location
Chr. 9: 83834651 - 83872390 on Build GRCm38
UniGene ID
Mm.1904
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001110265.1 NP_001103735.1 3 343
NM_001284272.1 NP_001271201.1 3 343
NM_009445.2 NP_033471.2 3 343
GenBank mRNA AK041487.1 - 3 314
AK049200.1 - 3 338
AK164093.1 - 3 342
AX775731.1 - 3 309
BC058851.1 - 3 329 30023533
BY729362.1 - 3 343
M86377.1 - 3 291

View Full Product Details