Gene Transcripts

Gene Symbol
Ulk1
Entrez Gene ID
22241
Gene Name
unc-51 like kinase 1
Gene Aliases
AU041434, Unc51.1, mKIAA0722
Chromosome Location
Chr. 5: 110784489 - 110810118 on Build GRCm38
UniGene ID
Mm.271898
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009469.3 NP_033495.2 20 2275
GenBank mRNA AF053756.1 - 20 1967
AF072370.1 - 20 1958
AK122357.1 - 19 1777
AK148317.1 - 20 2268
AK154939.1 - 20 2292
AK156534.1 - 20 2252
BC053062.1 - 20 1935
BC057121.1 - 20 2275 6834534
BC059835.1 - 20 2244 6406755

View Full Product Details