Gene Transcripts

Gene Symbol
Upk2
Entrez Gene ID
22269
Gene Name
uroplakin 2
Gene Aliases
AW491716, UPII
Chromosome Location
Chr. 9: 44452715 - 44454767 on Build GRCm38
UniGene ID
Mm.4453
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009476.2 NP_033502.2 3 393
GenBank mRNA BC104260.1 - 3 393
BC104261.1 - 3 393 40039814
DQ358978.1 - 3 332
U08030.1 - 3 391

View Full Product Details