Gene Transcripts

Gene Symbol
Xpc
Entrez Gene ID
22591
Gene Name
xeroderma pigmentosum, complementation group C
Gene Aliases
Chromosome Location
Chr. 6: 91489305 - 91515888 on Build GRCm38
UniGene ID
Mm.2806
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_009531.2 NP_033557.2 9 1152
GenBank mRNA AB071144.1 - 9 1089
AK004713.1 - 9 1092
AK028595.1 - 9 1090
AK166981.1 - 9 1087
AK170975.1 - 9 1084
BC052725.1 - 9 1114 6812198
U27398.1 - 9 1060
U40005.1 - 8 891

View Full Product Details