Gene Transcripts
- Gene Symbol
- Zfp60
- Entrez Gene ID
- 22718
- Gene Name
- zinc finger protein 60
- Gene Aliases
- 6330516O17Rik, AI426106, AI449354, Mfg-3, Mfg3
- Chromosome Location
- Chr. 7: 27731409 - 27751689 on Build GRCm38
- UniGene ID
- Mm.348021
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M28515.1 |
- |
1 |
329 |
|
View Full Product Details