Gene Transcripts

Gene Symbol
RAB35
Entrez Gene ID
11021
Gene Name
RAB35, member RAS oncogene family
Gene Aliases
H-ray, RAB1C, RAY
Chromosome Location
Chr. 12: 120095094 - 120116839 on Build GRCh38
UniGene ID
Hs.524788
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006861.6 NP_006852.1 5 581
XM_011537779.1 XP_011536081.1 5 518
GenBank mRNA AF498960.1 - 5 392
AK125543.1 - 6 1356
AK312538.1 - 5 491
BC015931.2 - 5 496 3907209
BM467132.1 - 5 563
BQ233709.1 - 5 537
BT020024.1 - 5 392
CR536486.1 - 5 392
CR541683.1 - 5 392
X79781.1 - 5 473

View Full Product Details