Gene Transcripts
- Gene Symbol
- RBPMS
- Entrez Gene ID
- 11030
- Gene Name
- RNA binding protein with multiple splicing
- Gene Aliases
- HERMES
- Chromosome Location
- Chr. 8: 30384501 - 30572261 on Build GRCh38
- UniGene ID
- Hs.334587
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001008710.2 |
NP_001008710.1
|
2, 3 |
744 |
|
NM_001008711.2 |
NP_001008711.1
|
2, 3 |
744 |
|
NM_001008712.2 |
NP_001008712.1
|
2, 3 |
744 |
|
NM_006867.3 |
NP_006858.1
|
2, 3 |
744 |
|
XM_017012980.1 |
XP_016868469.1
|
2, 3 |
739 |
|
XM_017012981.1 |
XP_016868470.1
|
2, 3 |
739 |
|
XM_017012982.1 |
XP_016868471.1
|
2, 3 |
739 |
|
XM_017012983.1 |
XP_016868472.1
|
2, 3 |
1725 |
|
XM_017012984.1 |
XP_016868473.1
|
2, 3 |
739 |
|
XM_017012986.1 |
XP_016868475.1
|
2, 3 |
739 |
|
XM_017012987.1 |
XP_016868476.1
|
2, 3 |
739 |
|
XM_017012988.1 |
XP_016868477.1
|
2, 3 |
739 |
|
XM_017012989.1 |
XP_016868478.1
|
2, 3 |
739 |
|
XM_017012990.1 |
XP_016868479.1
|
2, 3 |
739 |
|
XM_017012991.1 |
XP_016868480.1
|
2, 3 |
832 |
|
XM_017012992.1 |
XP_016868481.1
|
2, 3 |
739 |
|
GenBank mRNA |
AK304858.1 |
- |
3, 4 |
562 |
|
AK310533.1 |
- |
2, 3 |
172 |
|
BC003608.2 |
- |
2, 3 |
685 |
3505124 |
BC063494.1 |
- |
2, 3 |
151 |
|
BC092476.1 |
- |
2, 3 |
232 |
4837343 |
D84107.1 |
- |
2, 3 |
718 |
|
D84108.1 |
- |
2, 3 |
718 |
|
D84109.1 |
- |
2, 3 |
718 |
|
D84110.1 |
- |
2, 3 |
718 |
|
D84111.1 |
- |
1 |
18 |
|
View Full Product Details