Gene Transcripts

Gene Symbol
Spry1
Entrez Gene ID
24063
Gene Name
sprouty homolog 1 (Drosophila)
Gene Aliases
sprouty1, spry-1
Chromosome Location
Chr. 3: 37639846 - 37644599 on Build GRCm38
UniGene ID
Mm.330986
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
GenBank mRNA AF176903.1 - 3 2397
AK014578.1 - 3 2415
AK140405.1 - 3 2409
BC039139.1 - 4 2256 3672353
BC051977.1 - 2 2702
BC053428.1 - 3 2388 6506766
BC069914.1 - 1 1919 5251876

View Full Product Details