Gene Transcripts

Gene Symbol
Vat1
Entrez Gene ID
26949
Gene Name
vesicle amine transport protein 1 homolog (T californica)
Gene Aliases
VAT-1
Chromosome Location
Chr. 11: 101458748 - 101466199 on Build GRCm38
UniGene ID
Mm.29210
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012037.2 NP_036167.1 2 637
GenBank mRNA AK034795.1 - 2 639
AK145193.1 - 2 671
AK154178.1 - 622
AK154965.1 - 2 620
AK155342.1 - 2 638
AK159310.1 - 2 580
AK159315.1 - 2 622
AK160891.1 - 2 629
AK170371.1 - 2 618
AK181346.1 - 2 192
AK199541.1 - 1 118
BC025496.1 - 2 593 5320239
BC036169.1 - 1 79
BC085504.1 - 2 556
BC099678.1 - 2 619 6311681
X95562.1 - 2 259

View Full Product Details