Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001277101.1 | NP_001264030.1 | 2 | 409 | |
NM_013715.2 | NP_038743.1 | 2 | 658 | ||
GenBank mRNA | AF065386.1 | - | 2 | 411 | |
AF068223.1 | - | 2 | 426 | ||
AK002761.1 | - | 2 | 440 | ||
AK005728.1 | - | 2 | 445 | ||
AK012499.1 | - | 2 | 445 | ||
AK133073.1 | - | 2 | 438 | ||
AK146271.1 | - | 2 | 445 | ||
AK151492.1 | - | 2 | 444 | ||
BC046753.1 | - | 2 | 404 | 5693100 | |
CX240457.1 | - | 2 | 626 | ||
DV056728.1 | - | 2 | 465 | ||
U70736.1 | - | 2 | 434 |