Gene Transcripts
- Gene Symbol
- Dbf4
- Entrez Gene ID
- 27214
- Gene Name
- DBF4 homolog (S. cerevisiae)
- Gene Aliases
- AA545217, Ask
- Chromosome Location
- Chr. 5: 8396969 - 8422722 on Build GRCm38
- UniGene ID
- Mm.292470
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001190717.1 |
NP_001177646.1
|
9, 10 |
1013 |
|
NM_013726.3 |
NP_038754.1
|
9, 10 |
1016 |
|
GenBank mRNA |
AJ003132.1 |
- |
9, 10 |
906 |
|
AK014480.1 |
- |
9, 10 |
982 |
|
AK083650.1 |
- |
6, 7 |
391 |
|
AK132403.1 |
- |
9, 10 |
978 |
|
AK163408.1 |
- |
7, 8 |
592 |
|
BC069983.1 |
- |
9, 10 |
964 |
|
BC138709.1 |
- |
9, 10 |
834 |
8861731 |
BC138710.1 |
- |
9, 10 |
834 |
8861732 |
BC145392.1 |
- |
9, 10 |
831 |
9053950 |
View Full Product Details