Gene Transcripts

Gene Symbol
LILRA1
Entrez Gene ID
11024
Gene Name
leukocyte immunoglobulin like receptor A1
Gene Aliases
CD85I, LIR-6, LIR6
Chromosome Location
Chr. 19: 54593576 - 54602381 on Build GRCh38
UniGene ID
Hs.710507
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001278318.1 NP_001265247.1 8 1218
NM_006863.3 NP_006854.1 10 1818
NR_103501.1 9 1620
GenBank mRNA AF025529.1 - 8 1134
AF025530.1 - 10 1746
AK290919.1 - 10 1762

View Full Product Details