Gene Transcripts

Gene Symbol
Plekhb1
Entrez Gene ID
27276
Gene Name
pleckstrin homology domain containing, family B (evectins) member 1
Gene Aliases
AI315628, PHR1, Phret1, evt-1
Chromosome Location
Chr. 7: 100643896 - 100662394 on Build GRCm38
UniGene ID
Mm.491216
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163182.1 NP_001156654.1 7 742
NM_001163183.1 NP_001156655.1 7 700
NM_001163184.1 NP_001156656.1 6 595
NM_001163185.1 NP_001156657.1 6 585
NM_001163186.1 NP_001156658.1 6 691
NM_001163187.1 NP_001156659.1 5 586
NM_001291336.1 NP_001278265.1 5 555
NM_013746.3 NP_038774.1 8 847
GenBank mRNA AF000272.1 - 8 818
AF071000.1 - 5 574
AF100613.1 - 6 679
AF101053.1 - 7 713
AK005102.1 - 6 550
AK141634.1 - 553
AK158172.1 - 6 586
AK164683.1 - 6 550
BC024756.1 - 8 808 5369461
CX201870.1 - 7 652

View Full Product Details