Gene Transcripts

Gene Symbol
Sgsh
Entrez Gene ID
27029
Gene Name
N-sulfoglucosamine sulfohydrolase (sulfamidase)
Gene Aliases
4632406A19Rik
Chromosome Location
Chr. 11: 119343489 - 119355510 on Build GRCm38
UniGene ID
Mm.103788
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018822.3 NP_061292.2 2 204
GenBank mRNA AF153827.1 - 2 178
AF304053.1 - 2 178
AF304054.1 - 2 178
AK052266.1 - 2 197
AK076267.1 - 2 233
AK128926.1 - 2 204
AK141266.1 - 2 152
BC130268.1 - 2 201 40143074

View Full Product Details