Gene Transcripts
- Gene Symbol
- Tnfrsf4
- Entrez Gene ID
- 25572
- Gene Name
- tumor necrosis factor receptor superfamily, member 4
- Gene Aliases
- Ox40, Txgp1
- Chromosome Location
- Chr. 5: 173447784 - 173450474 on Build Rnor_6.0
- UniGene ID
- Rn.48883
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_013049.1 |
NP_037181.1
|
1 |
91 |
|
GenBank mRNA |
X17037.1 |
- |
1 |
91 |
|
View Full Product Details