Gene Transcripts

Gene Symbol
Pla2g5
Entrez Gene ID
29354
Gene Name
phospholipase A2, group V
Gene Aliases
Chromosome Location
Chr. 5: 157247608 - 157288716 on Build Rnor_6.0
UniGene ID
Rn.20244
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_017174.1 NP_058870.1 5 601
XM_006239139.2 XP_006239201.1 6 1400
XM_006239140.2 XP_006239202.1 6 1979
XM_006239141.2 XP_006239203.1 5 523
XM_008764209.1 XP_008762431.1 7 1514
XM_008764210.1 XP_008762432.1 5 2621
XM_008764211.1 XP_008762433.1 4 636
GenBank mRNA AY651028.1 - 5 468
BC085745.1 - 5 626 7192819
U03763.1 - 5 601

View Full Product Details