Gene Transcripts
- Gene Symbol
- Fgf5
- Entrez Gene ID
- 60662
- Gene Name
- fibroblast growth factor 5
- Gene Aliases
- Chromosome Location
- Chr. 14: 12974921 - 12995818 on Build Rnor_6.0
- UniGene ID
- Rn.44445
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022211.1 |
NP_071547.1
|
1, 2 |
341 |
|
GenBank mRNA |
D64085.1 |
- |
1, 2 |
341 |
|
View Full Product Details