Gene Transcripts
- Gene Symbol
- Masp1
- Entrez Gene ID
- 64023
- Gene Name
- mannan-binding lectin serine peptidase 1
- Gene Aliases
- Masp1/3, Masp3
- Chromosome Location
- Chr. 11: 80736425 - 80806278 on Build Rnor_6.0
- UniGene ID
- Rn.203236
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_022257.1 |
NP_071593.1
|
16 |
3710 |
|
GenBank mRNA |
AJ457084.1 |
- |
16 |
3710 |
|
View Full Product Details