Gene Transcripts

Gene Symbol
CEACAM7
Entrez Gene ID
1087
Gene Name
carcinoembryonic antigen related cell adhesion molecule 7
Gene Aliases
CGM2
Chromosome Location
Chr. 19: 41673307 - 41688277 on Build GRCh38
UniGene ID
Hs.74466
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001291485.1 NP_001278414.1 5 1928
NM_006890.4 NP_008821.2 5 1948
GenBank mRNA X98311.1 - 5 1838

View Full Product Details