Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001034009.1 | NP_001029181.1 | 6 | 640 | |
NM_023983.3 | NP_076473.2 | 6 | 640 | ||
GenBank mRNA | AB035506.1 | - | 6 | 638 | |
AB035507.1 | - | 6 | 638 | ||
BC070916.1 | - | 6 | 640 | 7105099 |