Gene Transcripts
- Gene Symbol
- Gucy2e
- Entrez Gene ID
- 79222
- Gene Name
- guanylate cyclase 2E
- Gene Aliases
- Chromosome Location
- Chr. 10: 55835695 - 55851235 on Build Rnor_6.0
- UniGene ID
- Rn.10060
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_024380.1 |
NP_077356.1
|
4 |
1128 |
|
GenBank mRNA |
L36029.1 |
- |
4 |
1128 |
|
View Full Product Details