Gene Transcripts

Gene Symbol
Nfkbib
Entrez Gene ID
81525
Gene Name
nuclear factor of kappa light polypeptide gene enhancer in B-cells inhibitor, beta
Gene Aliases
Chromosome Location
Chr. 1: 86941073 - 86948845 on Build Rnor_6.0
UniGene ID
Rn.8395
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_030867.2 NP_110494.2 3 726
GenBank mRNA AF246634.1 - 3 549
BC085729.1 - 3 726 7114624
FQ228042.1 - 3 523
FQ235103.1 - 3 671

View Full Product Details