Gene Transcripts

Gene Symbol
Dnmt1
Entrez Gene ID
84350
Gene Name
DNA (cytosine-5-)-methyltransferase 1
Gene Aliases
Chromosome Location
Chr. 8: 21922515 - 21968495 on Build Rnor_6.0
UniGene ID
Rn.6955
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_053354.3 NP_445806.3 3 268
GenBank mRNA AB012214.1 - 3 252
AF116344.1 - 3 306
D64060.1 - 3 140
DV214494.1 - 3 234

View Full Product Details